Gene Transcripts

Gene Symbol
Irgc1
Entrez Gene ID
210145
Gene Name
immunity-related GTPase family, cinema 1
Gene Aliases
F630044M05Rik, Gm1102, Gm474, Ifgge, Iigp5, Irgc
Chromosome Location
Chr. 7: 24431919 - 24445682 on Build GRCm38
UniGene ID
Mm.33811
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199013.2 NP_950178.2 3 1230
GenBank mRNA AK089224.1 - 3 1234
AK133005.1 - 2 1132

View Full Product Details