Gene Transcripts

Gene Symbol
Rhbdf2
Entrez Gene ID
217344
Gene Name
rhomboid 5 homolog 2 (Drosophila)
Gene Aliases
4732465I17Rik, Rhbdl6, cub
Chromosome Location
Chr. 11: 116598165 - 116627019 on Build GRCm38
UniGene ID
Mm.89695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167680.1 NP_001161152.1 17 2187
NM_172572.3 NP_766160.2 17 2401
GenBank mRNA AK028872.1 - 17 2402
AK154639.1 - 17 2168
AK171085.1 - 17 2183
AY167043.1 - 17 2187
BC031398.1 - 11 1220
BC052182.1 - 17 2135 6335022

View Full Product Details