Gene Transcripts

Gene Symbol
Mier3
Entrez Gene ID
218613
Gene Name
mesoderm induction early response 1, family member 3
Gene Aliases
5730509D14, AU015422, D130064H19Rik
Chromosome Location
Chr. 13: 111686157 - 111718596 on Build GRCm38
UniGene ID
Mm.31012
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172593.3 NP_766181.2 3 322
GenBank mRNA AK030747.1 - 3 250
AK083929.1 - 2 161
AK147427.1 - 3 323
BC138127.1 - 2 142 8861146

View Full Product Details