Gene Transcripts
- Gene Symbol
- Slc4a5
- Entrez Gene ID
- 232156
- Gene Name
- solute carrier family 4, sodium bicarbonate cotransporter, member 5
- Gene Aliases
- C330006M16, C330016K18Rik
- Chromosome Location
- Chr. 6: 83207580 - 83304945 on Build GRCm38
- UniGene ID
- Mm.333189
- Species
- Mus musculus
View Full Product Details