Gene Transcripts

Gene Symbol
Scgb2b24
Entrez Gene ID
233090
Gene Name
secretoglobin, family 2B, member 24
Gene Aliases
Abpbg24, Abpz, C2b, Scgb2b3
Chromosome Location
Chr. 7: 33737193 - 33739295 on Build GRCm38
UniGene ID
Mm.261648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177446.2 NP_803229.1 2 177
GenBank mRNA AA794526.1 - 2 177
AY370635.1 - 2 194
BC115602.1 - 2 177 40102624
BC115603.1 - 2 177 40102625
GU269236.1 - 2 167

View Full Product Details