Gene Transcripts

Gene Symbol
Samd15
Entrez Gene ID
238333
Gene Name
sterile alpha motif domain containing 15
Gene Aliases
Gm263
Chromosome Location
Chr. 12: 87196168 - 87213541 on Build GRCm38
UniGene ID
Mm.302304
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290288.1 NP_001277217.1 1 1159

View Full Product Details