Gene Transcripts

Gene Symbol
Ttc38
Entrez Gene ID
239570
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
9030019H09
Chromosome Location
Chr. 15: 85832304 - 85858822 on Build GRCm38
UniGene ID
Mm.151748
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033337.4 NP_001028509.2 13 1376
GenBank mRNA AK078848.1 - 13 1338
AK133636.1 - 12 1084
BC024550.1 - 13 1326
BC030849.1 - 13 1339
BC132621.1 - 13 1361 40130898
BC132623.1 - 13 1361 40130900

View Full Product Details