Gene Transcripts

Gene Symbol
Olfr93
Entrez Gene ID
258051
Gene Name
olfactory receptor 93
Gene Aliases
MOR256-39P
Chromosome Location
Chr. 17: 37151032 - 37151970 on Build GRCm38
UniGene ID
Mm.233576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011813.1 NP_001011813.1 1 920
GenBank mRNA BC139238.1 - 1 1001
BC139240.1 - 1 1001 8862262

View Full Product Details