Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012032.4 | NP_036162.3 | 8 | 1091 | |
GenBank mRNA | AB029499.1 | - | 8 | 1000 | |
AF181684.1 | - | 8 | 1019 | ||
AK031101.1 | - | 8 | 1080 | ||
AK128955.1 | - | 6 | 628 | ||
AK145604.1 | - | 8 | 1039 | ||
AK150106.1 | - | 8 | 1037 | ||
AK152720.1 | - | 8 | 1037 | ||
AK155921.1 | - | 8 | 1037 | ||
AK159347.1 | - | 8 | 1037 | ||
AK159488.1 | - | 8 | 1037 | ||
AK159819.1 | - | 8 | 1037 | ||
AK160005.1 | - | 8 | 1037 | ||
AK160055.1 | - | 8 | 1037 | ||
AK161981.1 | - | 8 | 1037 | ||
AK164807.1 | - | 8 | 1037 | ||
AK166924.1 | - | 8 | 1037 | ||
AK170350.1 | - | 8 | 1030 | ||
BC029980.1 | - | 8 | 1060 | ||
L29441.1 | - | 7 | 816 |