Gene Transcripts

Gene Symbol
Plch2
Entrez Gene ID
269615
Gene Name
phospholipase C, eta 2
Gene Aliases
A930027K05Rik, PLCeta2, Plc-eta2, Plcl4
Chromosome Location
Chr. 4: 154983115 - 155064091 on Build GRCm38
UniGene ID
Mm.379458
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113360.2 NP_001106831.1 7 1220
NM_175556.4 NP_780765.2 7 1220
GenBank mRNA AK044619.1 - 7 1219
AK049970.1 - 7 1064
AK078731.1 - 7 991
AK129147.1 - 7 980
AY966876.1 - 7 998
DQ176851.1 - 7 1220

View Full Product Details