Gene Transcripts

Gene Symbol
Zar1
Entrez Gene ID
317755
Gene Name
zygote arrest 1
Gene Aliases
Chromosome Location
Chr. 5: 72577113 - 72581361 on Build GRCm38
UniGene ID
Mm.11934
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174877.3 NP_777366.1 1, 2 794
GenBank mRNA AY191415.1 - 1, 2 794
BC099399.1 - 1, 2 788 30939000

View Full Product Details