Gene Transcripts

Gene Symbol
Dhps
Entrez Gene ID
330817
Gene Name
deoxyhypusine synthase
Gene Aliases
Dhs
Chromosome Location
Chr. 8: 85069718 - 85075162 on Build GRCm38
UniGene ID
Mm.193086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039514.1 NP_001034603.1 1 185
GenBank mRNA AK082081.1 - 1 196
AK159104.1 - 1 186
BC061234.1 - 1 146

View Full Product Details