Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004062.2 | NP_001004062.1 | 3 | 306 | |
GenBank mRNA | AK129173.1 | - | 3 | 303 | |
BC056137.1 | - | 3 | 312 | ||
BC068139.1 | - | 3 | 312 | ||
BC080308.1 | - | 3 | 306 | 5719562 |