Gene Transcripts

Gene Symbol
Omt2b
Entrez Gene ID
382088
Gene Name
oocyte maturation, beta
Gene Aliases
AU018149, C86230, Mosg, OM2b
Chromosome Location
Chr. 9: 78328030 - 78329620 on Build GRCm38
UniGene ID
Mm.360477
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205822.2 NP_991391.2 2 280
GenBank mRNA BC115754.1 - 1 513
CO795725.1 - 2 171
S81935.1 - 3 296

View Full Product Details