Gene Transcripts

Gene Symbol
Arhgef15
Entrez Gene ID
442801
Gene Name
Rho guanine nucleotide exchange factor (GEF) 15
Gene Aliases
D130071N09, D530030K12Rik, E5
Chromosome Location
Chr. 11: 68927611 - 68956934 on Build GRCm38
UniGene ID
Mm.376147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177566.3 NP_808234.2 12 2181
GenBank mRNA AK052570.1 - 5 521
AK173061.1 - 8 886
BC089365.1 - 12 2181 6844583
BC116343.1 - 12 2157 40098168
BC116344.1 - 12 2157 40098169

View Full Product Details