Gene Transcripts

Gene Symbol
Rnase13
Entrez Gene ID
497071
Gene Name
ribonuclease, RNase A family, 13 (non-active)
Gene Aliases
Chromosome Location
Chr. 14: 51921365 - 51923277 on Build GRCm38
UniGene ID
Mm.378602
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011687.2 NP_001011687.1 1 520
GenBank mRNA BC132614.1 - 1 520
BC132616.1 - 1 520 40130893

View Full Product Details