Gene Transcripts

Gene Symbol
Rgs11
Entrez Gene ID
50782
Gene Name
regulator of G-protein signaling 11
Gene Aliases
C78048
Chromosome Location
Chr. 17: 26202962 - 26211324 on Build GRCm38
UniGene ID
Mm.109840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081069.1 NP_001074538.1 16 1266
GenBank mRNA AF061934.1 - 15 1171
AK039314.1 - 15 1888
AK080707.1 - 16 1360
AK161551.1 - 12 1325
AK166716.1 - 2 511
BC019741.1 - 5 598
BC049910.1 - 14 1970

View Full Product Details