Gene Transcripts

Gene Symbol
Cdk2ap2
Entrez Gene ID
52004
Gene Name
CDK2-associated protein 2
Gene Aliases
5830466O21Rik, D19Ertd144e, Doc-1r
Chromosome Location
Chr. 19: 4096843 - 4099019 on Build GRCm38
UniGene ID
Mm.29609
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026373.4 NP_080649.1 4 427
GenBank mRNA AK003686.2 - 4 393
AK012121.1 - 4 430
AK018035.1 - 4 427
BC025656.1 - 4 393 5342719

View Full Product Details