Gene Transcripts

Gene Symbol
Ldlrad1
Entrez Gene ID
546840
Gene Name
low density lipoprotein receptor class A domain containing 1
Gene Aliases
OTTMUSG00000008594
Chromosome Location
Chr. 4: 107209078 - 107221101 on Build GRCm38
UniGene ID
Mm.331755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081272.1 NP_001074741.1 1 306

View Full Product Details