Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019662.2 | NP_062636.2 | 2, 3 | 641 | |
GenBank mRNA | AF084466.1 | - | 1, 2 | 384 | |
AK077271.1 | - | 2, 3 | 639 | ||
AK087473.1 | - | 2, 3 | 614 | ||
AK159550.1 | - | 1, 2 | 424 | ||
BC057138.1 | - | 2, 3 | 620 | 5149047 |