Gene Transcripts
- Gene Symbol
- Dnajb8
- Entrez Gene ID
- 56691
- Gene Name
- DnaJ (Hsp40) homolog, subfamily B, member 8
- Gene Aliases
- 1700016F14Rik, 1700031F10Rik, AI326313, mDj6
- Chromosome Location
- Chr. 6: 88222268 - 88223256 on Build GRCm38
- UniGene ID
- Mm.272871
- Species
- Mus musculus
- Gene Symbol
- 1700031F10Rik
- Entrez Gene ID
- 73317
- Gene Name
- RIKEN cDNA 1700031F10 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.272871
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006579.1 |
- |
2 |
516 |
|
BC099573.1 |
- |
2 |
574 |
|
View Full Product Details