Gene Transcripts

Gene Symbol
Icmt
Entrez Gene ID
57295
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
1700008E11Rik, C80758, Gm13095, HSTE14, OTTMUSG00000010406, PPMT, STE14, pcCMT
Chromosome Location
Chr. 4: 152297214 - 152307126 on Build GRCm38
UniGene ID
Mm.277464
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133788.2 NP_598549.1 4, 5 833
GenBank mRNA AF209926.1 - 4, 5 615
AK154135.1 - 4, 5 765
AK154410.1 - 5, 6 856
BC057132.1 - 5, 6 852
BC110695.1 - 4, 5 820 6842587

View Full Product Details