Gene Transcripts

Gene Symbol
Sectm1b
Entrez Gene ID
58210
Gene Name
secreted and transmembrane 1B
Gene Aliases
1810003C24Rik, K12, Sectm1
Chromosome Location
Chr. 11: 121053423 - 121063569 on Build GRCm38
UniGene ID
Mm.116986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026907.3 NP_081183.3 5 762
GenBank mRNA AF210700.1 - 4 665
AK007321.1 - 4 624
AK029082.1 - 5 763
AK143807.1 - 5 748
BC010805.1 - 5 839 4239879
BC020159.1 - 4 710 4016267

View Full Product Details