Gene Transcripts
- Gene Symbol
- Nek6
- Entrez Gene ID
- 59126
- Gene Name
- NIMA (never in mitosis gene a)-related expressed kinase 6
- Gene Aliases
- 1300007C09Rik
- Chromosome Location
- Chr. 2: 38511697 - 38587490 on Build GRCm38
- UniGene ID
- Mm.143818
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159631.1 |
NP_001153103.1
|
5, 6 |
528 |
|
NM_021606.3 |
NP_067619.1
|
5, 6 |
521 |
|
GenBank mRNA |
AF218847.1 |
- |
5, 6 |
523 |
|
AK004925.1 |
- |
5, 6 |
498 |
|
AK011519.1 |
- |
5, 6 |
507 |
|
AK018168.1 |
- |
5, 6 |
511 |
|
AK029266.1 |
- |
5, 6 |
510 |
|
AK075717.1 |
- |
5, 6 |
495 |
|
AK075836.1 |
- |
5, 6 |
507 |
|
AK086700.1 |
- |
|
526 |
|
AK089919.1 |
- |
5, 6 |
482 |
|
AK170757.1 |
- |
5, 6 |
528 |
|
BC019524.1 |
- |
5, 6 |
590 |
4209097 |
View Full Product Details