Gene Transcripts
- Gene Symbol
- Tmem14c
- Entrez Gene ID
- 66154
- Gene Name
- transmembrane protein 14C
- Gene Aliases
- 1110021D01Rik, AU020193, HSPC194
- Chromosome Location
- Chr. 13: 41016250 - 41022582 on Build GRCm38
- UniGene ID
- Mm.30005
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025387.3 |
NP_079663.1
|
2, 3 |
207 |
|
GenBank mRNA |
AK003891.1 |
- |
2, 3 |
156 |
|
AK008878.1 |
- |
2, 3 |
129 |
|
AK010609.1 |
- |
2, 3 |
167 |
|
AK012101.1 |
- |
2, 3 |
154 |
|
AK019395.1 |
- |
2, 3 |
156 |
|
AK050688.1 |
- |
2, 3 |
142 |
|
BC025854.1 |
- |
2, 3 |
108 |
5250659 |
BC055862.1 |
- |
2, 3 |
131 |
3492090 |
BC069899.1 |
- |
1, 2 |
853 |
|
BY785978.1 |
- |
2, 3 |
207 |
|
View Full Product Details