Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023179.3 | NP_075668.1 | 3 | 470 | |
GenBank mRNA | AB076405.1 | - | 3 | 525 | |
AK005109.1 | - | 3 | 350 | ||
AK011945.1 | - | 3 | 313 | ||
BC020190.1 | - | 3 | 466 | 4505711 | |
BC025190.1 | - | 866 | |||
BC049587.1 | - | 3 | 335 | ||
BC062380.1 | - | 3 | 458 | 6468413 | |
BU558667.1 | - | 3 | 473 |