Gene Transcripts

Gene Symbol
Susd3
Entrez Gene ID
66329
Gene Name
sushi domain containing 3
Gene Aliases
1700017I11Rik, 2810440J20Rik
Chromosome Location
Chr. 13: 49230827 - 49248706 on Build GRCm38
UniGene ID
Mm.34308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025491.3 NP_079767.2 4 531
NM_028340.1 NP_082616.1 4 700
GenBank mRNA AK003860.1 - 4 531
AK006068.1 - 4 521
BC045525.1 - 4 700 6468669

View Full Product Details