Gene Transcripts

Gene Symbol
Hspbap1
Entrez Gene ID
66667
Gene Name
Hspb associated protein 1
Gene Aliases
3830421G21Rik
Chromosome Location
Chr. 16: 35769663 - 35828462 on Build GRCm38
UniGene ID
Mm.45272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175111.3 NP_780320.2 8 1436
GenBank mRNA AK017731.1 - 8 1391
AK028824.1 - 9 1497
AK076189.1 - 8 1403
AK089941.1 - 8 1452

View Full Product Details