Gene Transcripts
- Gene Symbol
- Tctex1d1
- Entrez Gene ID
- 67344
- Gene Name
- Tctex1 domain containing 1
- Gene Aliases
- 1700055O19Rik
- Chromosome Location
- Chr. 4: 102978388 - 103005594 on Build GRCm38
- UniGene ID
- Mm.111346
- Species
- Mus musculus
- Gene Symbol
- BB031773
- Entrez Gene ID
- 100473
- Gene Name
- expressed sequence BB031773
- Gene Aliases
- Chromosome Location
- Chr. 4: 103004106 - 103011965 on Build GRCm38
- UniGene ID
- Mm.393171
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK030945.1 |
- |
4 |
3591 |
|
View Full Product Details