Gene Transcripts

Gene Symbol
Yipf4
Entrez Gene ID
67864
Gene Name
Yip1 domain family, member 4
Gene Aliases
2310034L04Rik
Chromosome Location
Chr. 17: 74489493 - 74500277 on Build GRCm38
UniGene ID
Mm.41891
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026417.4 NP_080693.2 6 854
GenBank mRNA AK017721.1 - 6 833
AK017722.1 - 6 833
AK083301.1 - 6 834
AK159227.1 - 6 815
AK166347.1 - 6 842
BC003936.1 - 7 1029 3486503

View Full Product Details