Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039483.1 | NP_001034572.1 | 7 | 741 | |
GenBank mRNA | AK004504.1 | - | 7 | 583 | |
AK015068.1 | - | 7 | 1032 | ||
AK037709.1 | - | 7 | 676 | ||
BC011457.1 | - | 7 | 615 | 3585288 | |
BC019761.1 | - | 7 | 659 | 5009960 | |
BC020098.1 | - | 7 | 671 | 4019484 |