Gene Transcripts

Gene Symbol
Fbxl2
Entrez Gene ID
72179
Gene Name
F-box and leucine-rich repeat protein 2
Gene Aliases
2810423A21Rik, Fbl3
Chromosome Location
Chr. 9: 113958757 - 114026787 on Build GRCm38
UniGene ID
Mm.386808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178624.6 NP_848739.1 5 286
GenBank mRNA AK013142.1 - 6 420
AK039010.1 - 5 286
AK044693.1 - 5 283
AK045742.1 - 5 273
AK089994.1 - 5 262
AK141177.1 - 5 276
AK163472.1 - 5 282
BC096582.1 - 5 272 3963599
BC145665.1 - 5 266 40130968
BC145998.1 - 5 266 40131301

View Full Product Details