Gene Transcripts

Gene Symbol
Psma8
Entrez Gene ID
73677
Gene Name
proteasome (prosome, macropain) subunit, alpha type, 8
Gene Aliases
2410072D24Rik
Chromosome Location
Chr. 18: 14706135 - 14762301 on Build GRCm38
UniGene ID
Mm.87277
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163609.1 NP_001157081.1 6, 7 733
GenBank mRNA AK010717.1 - 6, 7 734
BC019368.1 - 3, 4 199
BC116990.1 - 6, 7 695 40126309
BC116992.1 - 6, 7 695 40126311
BC144771.1 - 7, 8 766 9053319
BC144772.1 - 5, 6 575 9053320

View Full Product Details