Gene Transcripts

Gene Symbol
Ppapdc2
Entrez Gene ID
74411
Gene Name
phosphatidic acid phosphatase type 2 domain containing 2
Gene Aliases
4932443D16Rik
Chromosome Location
Chr. 19: 28963920 - 28966801 on Build GRCm38
UniGene ID
Mm.441887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028922.3 NP_083198.1 1 570
GenBank mRNA AK016572.1 - 1 571
AK041581.1 - 1 553
AK149181.1 - 1 332
BC052412.1 - 1 537 5720146

View Full Product Details