Gene Transcripts

Gene Symbol
Fer1l4
Entrez Gene ID
74562
Gene Name
fer-1-like 4 (C. elegans)
Gene Aliases
9130402C12Rik
Chromosome Location
Chr. 2: 156019139 - 156053018 on Build GRCm38
UniGene ID
Mm.159241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136556.1 NP_001130028.1 38 4795
GenBank mRNA AK018656.1 - 2 372
AK035669.1 - 16 1937

View Full Product Details