Gene Transcripts

Gene Symbol
Rab11fip2
Entrez Gene ID
74998
Gene Name
RAB11 family interacting protein 2 (class I)
Gene Aliases
4930470G04Rik, A830046J09Rik, AW558126, Nrip11
Chromosome Location
Chr. 19: 59760441 - 59943666 on Build GRCm38
UniGene ID
Mm.24167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033172.3 NP_001028344.2 1 503
GenBank mRNA AK154916.1 - 1 503
BC139380.1 - 1 266 8862402
BC139419.1 - 1 266 8862441

View Full Product Details