Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029420.4 | NP_083696.2 | 1 | 311 | |
GenBank mRNA | AK010748.1 | - | 1 | 312 | |
AK010828.1 | - | 1 | 275 | ||
AK014753.1 | - | 1 | 168 | ||
AK049139.1 | - | 1 | 312 | ||
AK053901.1 | - | 1 | 252 | ||
BC055909.1 | - | 1 | 263 | ||
BC145468.1 | - | 1 | 175 | ||
BC145688.1 | - | 1 | 176 | 40130991 |