Gene Transcripts

Gene Symbol
Spcs3
Entrez Gene ID
76687
Gene Name
signal peptidase complex subunit 3 homolog (S. cerevisiae)
Gene Aliases
1810011E08Rik
Chromosome Location
Chr. 8: 54520433 - 54530739 on Build GRCm38
UniGene ID
Mm.86437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029701.1 NP_083977.1 4, 5 506
GenBank mRNA AK007432.1 - 4, 5 485
AK028314.1 - 4, 5 468
AK048051.1 - 4, 5 492
BC019390.1 - 4, 5 445
BC054817.1 - 4, 5 471 5375997

View Full Product Details