Gene Transcripts

Gene Symbol
Ston1
Entrez Gene ID
77057
Gene Name
stonin 1
Gene Aliases
4921524J06Rik, C86231, Salf, Sblf
Chromosome Location
Chr. 17: 88597635 - 88645724 on Build GRCm38
UniGene ID
Mm.482203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029858.2 NP_084134.2 2 1158
GenBank mRNA AK014958.1 - 2 1093
AK029878.1 - 1105
AK029959.1 - 1138
AK077966.1 - 2 885
AK136565.1 - 2 1012

View Full Product Details