Gene Transcripts

Gene Symbol
Cypt15
Entrez Gene ID
78631
Gene Name
cysteine-rich perinuclear theca 15
Gene Aliases
1700085A12Rik
Chromosome Location
Chr. X: 39346266 - 39346963 on Build GRCm38
UniGene ID
Mm.243738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177380.1 NP_001170851.1 2 269
GenBank mRNA AK018908.1 - 2 269
BC100466.1 - 2 269

Gene Symbol
Cypt14
Entrez Gene ID
100504400
Gene Name
cysteine-rich perinuclear theca 14
Gene Aliases
Chromosome Location
Chr. X: 39862919 - 39863604 on Build GRCm38
UniGene ID
Mm.373409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191032.1 NP_001177961.1 2 269
GenBank mRNA BC099555.1 - 2 269

View Full Product Details