Gene Transcripts

Gene Symbol
Plekhj1
Entrez Gene ID
78670
Gene Name
pleckstrin homology domain containing, family J member 1
Gene Aliases
9530063M10Rik
Chromosome Location
Chr. 10: 80796099 - 80798626 on Build GRCm38
UniGene ID
Mm.41479
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023900.2 NP_076389.2 5 803
GenBank mRNA AK051734.1 - 5 460
AK088567.1 - 5 766
AK150144.1 - 5 418
AK178993.1 - 3 208
AK188371.1 - 1 39
AK219000.1 - 3 140
BC005565.1 - 5 401 3966305
BE951415.1 - 2 470

View Full Product Details