Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024281.2 | NP_077243.2 | 2 | 424 | |
NM_133626.2 | NP_598329.1 | 2 | 424 | ||
GenBank mRNA | AF273683.1 | - | 347 | ||
AF273684.1 | - | 2 | 347 | ||
AF273685.1 | - | 2 | 347 | ||
AF273686.1 | - | 2 | 347 | ||
AF273687.1 | - | 2 | 347 | ||
AF273688.1 | - | 2 | 347 | ||
AF273689.1 | - | 2 | 347 | ||
AF273690.1 | - | 2 | 347 | ||
AF273691.1 | - | 2 | 347 | ||
AF273692.1 | - | 2 | 347 | ||
AF273693.1 | - | 2 | 347 | ||
AK039114.1 | - | 2 | 424 | ||
BC002105.1 | - | 2 | 334 | ||
BC031452.1 | - | 2 | 413 |