Gene Transcripts

Gene Symbol
Cnnm1
Entrez Gene ID
83674
Gene Name
cyclin M1
Gene Aliases
AI846314, Acdp1, CLP-1
Chromosome Location
Chr. 19: 43440425 - 43497213 on Build GRCm38
UniGene ID
Mm.329864
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031396.2 NP_113573.2 1 1549
GenBank mRNA AF202994.1 - 1 775
DQ885890.5 - 1 445
EF041508.1 - 1 445

View Full Product Details