Product Details
- SNP ID
-
rs4159447
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:8585484 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTTCACCCTCTAGAGGAAGTCT[A/C]CCCTAGGTGATACTTGGTGTTTAAT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Abat
PubMed Links
Gene Details
- Gene
- Abat
- Gene Name
- 4-aminobutyrate aminotransferase
View Full Product Details