Product Details

SNP ID
rs6238987
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:140365996 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTAAAGCAGCCCTCCCTTCAGCCTC[A/G]CAGCCATAGAATGGAATCATGACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Sel1l2 PubMed Links

Gene Details

Gene
Sel1l2
Gene Name
sel-1 suppressor of lin-12-like 2 (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033296.2 Intron NP_001028468.1

View Full Product Details