Product Details
- SNP ID
-
rs29816714
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:141977504 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCTAGAGCAAGCAGGAACTGAG[C/T]GAACAGAGTTGACCAAAGGGAGCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Macrod2
PubMed Links
Gene Details
- Gene
- Macrod2
- Gene Name
- MACRO domain containing 2
View Full Product Details