Product Details

SNP ID
rs13476815
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:143881481 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCAAAGGCCAGTGGACACAGAATCC[A/G]TGTGATGCTGTACCCATCCTGGATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Bfsp1 PubMed Links

Gene Details

Gene
Bfsp1
Gene Name
beaded filament structural protein 1, in lens-CP94
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009751.2 Intron NP_033881.2

View Full Product Details