Product Details

SNP ID
rs6203474
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:157002514 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATGGTACTGGGGAGCCTCTCAGCTG[A/C]TCCGCTGGCAGCAGCAGAGCTCAGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Dsn1 PubMed Links

Gene Details

Gene
Dsn1
Gene Name
DSN1, MIND kinetochore complex component, homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025853.3 Intron NP_080129.2
Gene
Soga1
Gene Name
suppressor of glucose, autophagy associated 1
There are no transcripts associated with this gene.

View Full Product Details