Product Details
- SNP ID
-
rs3659869
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:132700832 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTACATCTTGGCAAACTGGGAAG[C/T]CATGTGCTCAGCCTATAAATCTCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tbck
PubMed Links
Gene Details
- Gene
- Tbck
- Gene Name
- TBC1 domain containing kinase
There are no transcripts associated with this gene.
View Full Product Details