Product Details

SNP ID
rs13480663
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:80431055 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTTGGAACTGGGGTGCGTCCCCCAG[A/G]TCTAGAGACAGCCCTGTGGTGACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Tcf3 PubMed Links

Gene Details

Gene
Tcf3
Gene Name
transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164147.1 348 UTR 5 NP_001157619.1
NM_001164148.1 348 UTR 5 NP_001157620.1
NM_001164149.1 348 UTR 5 NP_001157621.1
NM_001164150.1 348 UTR 5 NP_001157622.1
NM_001164151.1 348 UTR 5 NP_001157623.1
NM_001164152.1 348 UTR 5 NP_001157624.1
NM_001164153.1 348 UTR 5 NP_001157625.1
NM_011548.4 348 UTR 5 NP_035678.3

View Full Product Details